Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nck-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001375010.1 | NP_001361996.1 | 2 - 3 | 274 | 55 | |
NM_076305.6 | NP_508706.2 | 2 - 3 | 274 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Caenorhabditis elegans Probe spans exons |