Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vha-12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076310.8 | NP_508711.1 | 3 - 4 | 1338 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |