Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cka-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029309.5 | NP_001024480.1 | 6 - 7 | 1177 | 107 | |
NM_001029310.6 | NP_001024481.1 | 6 - 7 | 2292 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |