Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dop-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029407.4 | NP_001024578.1 | 7 - 8 | 1005 | 69 | |
NM_001029408.5 | NP_001024579.1 | 6 - 7 | 916 | 69 | ||
NM_001383561.1 | 8 - 9 | 1789 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |