Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029194.4 | NP_001024365.1 | 9 - 10 | 1339 | 107 | |
NM_001380949.1 | NP_001367445.1 | 10 - 11 | 1264 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |