Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: flp-12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076388.5 | NP_508789.1 | 1 - 2 | 138 | 64 | |
GenBank mRNA | AF042399.1 | - | 1 - 2 | 61 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |