Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: aqp-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029587.3 | NP_001024758.1 | 1 - 2 | 97 | 101 | |
NM_001373283.2 | NP_001359963.1 | 1 - 2 | 97 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |