Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mig-13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029489.6 | NP_001024660.1 | 7 - 8 | 909 | 69 | |
NM_001029490.5 | NP_001024661.1 | 5 - 6 | 795 | 69 | ||
GenBank mRNA | AF150958.1 | - | 6 - 7 | 813 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Caenorhabditis elegans Probe spans exons |