Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ztf-16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029679.5 | NP_001024850.1 | 4 - 5 | 377 | 75 | |
NM_001029680.5 | NP_001024851.1 | 4 - 5 | 383 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Caenorhabditis elegans Probe spans exons |