Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: add-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029468.5 | NP_001024639.1 | 14 - 15 | 1930 | 66 | |
NM_001029469.5 | NP_001024640.1 | 14 - 15 | 1937 | 66 | ||
NM_001029470.4 | NP_001024641.1 | 14 - 15 | 1939 | 66 | ||
NM_001380952.1 | NP_001367832.1 | 14 - 15 | 1930 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |