Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ifa-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076435.9 | NP_508836.5 | 5 - 6 | 878 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |