Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: set-30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076449.6 | NP_508850.2 | 1 - 2 | 164 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |