Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pqn-62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313445.3 | NP_001300374.1 | 1 - 2 | 624 | 65 | |
NM_001313446.4 | NP_001300375.1 | 3 - 4 | 921 | 65 | ||
NM_001380960.1 | NP_001368582.1 | 3 - 4 | 900 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |