Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pqn-62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313445.3 | NP_001300374.1 | 8 - 9 | 1871 | 66 | |
NM_001313446.4 | NP_001300375.1 | 10 - 11 | 2168 | 66 | ||
NM_001380960.1 | NP_001368582.1 | 10 - 11 | 2147 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |