Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vps-41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001380962.1 | NP_001367568.1 | 5 - 6 | 952 | 76 | |
NM_001383575.1 | 5 - 6 | 957 | 76 | |||
NM_001383576.1 | 3 - 4 | 526 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |