Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: acs-17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001373312.4 | NP_001360705.1 | 6 | 730 | 87 | |
NM_076592.9 | NP_508993.2 | 11 - 12 | 1842 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |