Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: inx-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076601.7 | NP_509002.2 | 1 - 2 | 155 | 90 | |
GenBank mRNA | U59211.1 | - | 1 - 2 | 155 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |