Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ggr-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029301.6 | NP_001024472.2 | 9 - 10 | 1118 | 91 | |
NM_001380988.1 | NP_001368738.1 | 8 - 9 | 980 | 91 | ||
NM_001380989.1 | NP_001368736.1 | 8 - 9 | 980 | 91 | ||
NM_001380990.1 | NP_001368737.1 | 8 - 9 | 980 | 91 | ||
NM_001380991.1 | NP_001368739.1 | 8 - 9 | 980 | 91 | ||
NM_001380992.2 | NP_001368740.1 | 8 - 9 | 1050 | 91 | ||
NM_001380999.1 | NP_001367470.1 | 8 - 9 | 980 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |