Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: prx-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129639.3 | NP_001123111.1 | 5 - 6 | 610 | 89 | |
NM_001383596.1 | 4 - 5 | 675 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |