Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mom-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076653.5 | NP_509054.1 | 7 - 8 | 1126 | 99 | |
GenBank mRNA | AF013489.1 | - | 7 - 8 | 1123 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Caenorhabditis elegans Probe spans exons |