Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: glr-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001381009.1 | NP_001368711.1 | 4 - 5 | 413 | 64 | |
NM_001381010.2 | NP_001367505.1 | 4 - 5 | 709 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |