Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ets-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001373331.1 | NP_001359862.1 | 7 - 8 | 850 | 60 | |
NM_076698.6 | NP_509099.2 | 4 - 5 | 440 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |