Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dop-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029736.6 | NP_001024907.2 | 8 - 9 | 1337 | 100 | |
NM_001029737.5 | NP_001024908.2 | 8 - 9 | 1141 | 100 | ||
NM_001038472.5 | NP_001033561.1 | 7 - 8 | 1520 | 100 | ||
NM_001381012.1 | NP_001368078.1 | 8 - 9 | 1103 | 100 | ||
GenBank mRNA | AY485309.1 | - | 8 - 9 | 1137 | 100 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor Detect Genomic DNA Caenorhabditis elegans Probe spans exons |