Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nas-11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029618.5 | NP_001024789.1 | 3 - 4 | 588 | 73 | |
NM_001029619.7 | NP_001024790.1 | 3 - 4 | 587 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |