Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nas-11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029618.5 | NP_001024789.1 | 4 - 5 | 663 | 102 | |
NM_001029619.7 | NP_001024790.1 | 4 - 5 | 662 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |