Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: abts-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029655.6 | NP_001024826.2 | 4 - 5 | 772 | 61 | |
NM_001129697.5 | NP_001123169.1 | 4 - 5 | 772 | 61 | ||
NM_001129698.4 | NP_001123170.1 | 4 - 5 | 730 | 61 | ||
NM_001373337.3 | NP_001360088.1 | 4 - 5 | 778 | 61 | ||
GenBank mRNA | AY887906.1 | - | 4 - 5 | 794 | 61 | |
AY954522.1 | - | 4 - 5 | 794 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |