Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: odr-10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076756.4 | NP_509157.1 | 2 - 3 | 363 | 83 | |
GenBank mRNA | U49449.1 | - | 2 - 3 | 368 | 83 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Detect Genomic DNA Caenorhabditis elegans Probe spans exons |