Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: odr-10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076756.4 | NP_509157.1 | 3 - 4 | 493 | 161 | |
GenBank mRNA | U49449.1 | - | 3 - 4 | 498 | 161 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Detect Genomic DNA Caenorhabditis elegans Probe spans exons |