Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076764.5 | NP_509165.1 | 12 - 13 | 1846 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |