Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076764.5 | NP_509165.1 | 5 - 6 | 1100 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Caenorhabditis elegans Probe spans exons |