Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tyra-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029351.6 | NP_001024522.1 | 5 - 6 | 665 | 97 | |
NM_001038448.5 | NP_001033537.1 | 5 - 6 | 673 | 97 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor Detect Genomic DNA Caenorhabditis elegans Probe spans exons |