Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ceh-14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076872.4 | NP_509273.1 | 3 - 4 | 302 | 71 | |
GenBank mRNA | AF244368.1 | - | 3 - 4 | 316 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Caenorhabditis elegans Probe spans exons |