Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: glb-16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076874.8 | NP_509275.2 | 3 - 4 | 308 | 85 | |
NR_132546.1 | 2 - 3 | 589 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |