Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: glb-16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076874.8 | NP_509275.2 | 6 - 7 | 921 | 63 | |
NR_132546.1 | 5 - 6 | 1202 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |