Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: apy-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076882.10 | NP_509283.1 | 8 - 9 | 2161 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |