Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dhs-29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076893.4 | NP_509294.1 | 1 - 2 | 118 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |