Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: stn-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029439.4 | NP_001024610.1 | 3 - 4 | 360 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |