Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029435.5 | NP_001024606.1 | 7 - 8 | 1386 | 88 | |
NR_136373.1 | 6 - 7 | 1400 | 88 | |||
GenBank mRNA | S66176.1 | - | 7 - 8 | 1350 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |