Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kvs-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029318.5 | NP_001024489.1 | 4 - 5 | 574 | 66 | |
NM_001029319.4 | NP_001024490.1 | 4 - 5 | 548 | 66 | ||
NM_001373368.2 | NP_001360715.1 | 4 - 5 | 578 | 66 | ||
NM_001375071.1 | NP_001361979.1 | 4 - 5 | 533 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |