Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ref-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029306.4 | NP_001024477.1 | 2 - 3 | 182 | 80 | |
NM_001029307.5 | NP_001024478.1 | 3 - 4 | 213 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |