Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ref-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029306.4 | NP_001024477.1 | 6 - 7 | 666 | 64 | |
NM_001029307.5 | NP_001024478.1 | 7 - 8 | 697 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Caenorhabditis elegans Probe spans exons |