Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: deg-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001373369.2 | NP_001360711.1 | 16 - 17 | 1986 | 72 | |
NM_001373370.2 | NP_001360706.1 | 16 | 2244 | 72 | ||
NM_001373371.2 | NP_001360707.1 | 15 | 2076 | 72 | ||
NM_001373372.2 | NP_001360708.1 | 14 | 2022 | 72 | ||
NM_001373373.2 | NP_001360709.1 | 15 | 2214 | 72 | ||
NM_001373374.2 | NP_001360710.1 | 13 | 1992 | 72 | ||
NM_076910.5 | NP_509311.1 | 14 - 15 | 2056 | 72 | ||
GenBank mRNA | L34414.1 | - | 14 - 15 | 2078 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |