Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zig-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076935.6 | NP_509336.1 | 2 - 3 | 494 | 70 | |
GenBank mRNA | AF456250.1 | - | 2 - 3 | 435 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |