Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: xol-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029247.3 | NP_001024418.1 | 3 - 4 | 368 | 68 | |
NM_001029248.1 | NP_001024419.1 | 3 - 4 | 354 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |