Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kqt-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076991.7 | NP_509392.3 | 12 - 13 | 1651 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |