Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kqt-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076991.7 | NP_509392.3 | 8 - 9 | 1215 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |