Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sulp-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077023.5 | NP_509424.2 | 2 - 3 | 320 | 72 | |
GenBank mRNA | AY887909.1 | - | 2 - 3 | 320 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |