Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ced-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077026.10 | NP_509427.2 | 7 - 8 | 1184 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |