Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nlp-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270133.3 | NP_001257062.1 | 3 - 4 | 206 | 109 | |
NM_001270134.3 | NP_001257063.1 | 2 - 3 | 114 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |