Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rig-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001375088.2 | NP_001362001.1 | 14 - 15 | 2577 | 138 | |
NM_001383616.1 | 13 - 14 | 2532 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons |